Invitrogen
Manufacturer Code:PA5102857
Catalog # PIPA5102857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CLCN3 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chloride channel 3; chloride channel 3 chloride channel protein 3 chloride channel voltage-sensitive 3 chloride transporter ClC-3 chloride voltage-gated channel 3 CIC3 CLC3 clC-3 CLCN 3 CLCN3 DKFZp564I0463 H(+)/Cl(-) exchange transporter 3 protein kinase C-regulated chloride channel; Chloride channel protein 3; chloride channel, voltage-sensitive 3; Chloride transporter ClC-3; ClC-3; H(+)/Cl(-) exchange transporter 3; protein kinase C-regulated chloride channel
Gene Aliases: ClC-3; CLC3; CLCN3
UniProt ID: (Human) P51790, (Rat) P51792, (Mouse) P51791
Entrez Gene ID: (Human) 1182, (Rat) 84360, (Mouse) 12725
Molecular Function: anion channel ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.