Novus Biologicals
Manufacturer Code:NBP287193
Catalog # NB034815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human CLCN3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chloride channel 3; chloride channel 3 Chloride channel protein 3 Chloride transporter ClC-3 ClC-3 CLC3 H(+)/Cl(-) exchange transporter 3; Chloride channel protein 3; chloride channel, voltage-sensitive 3; Chloride transporter ClC-3; ClC-3; H(+)/Cl(-) exchange transporter 3
Gene Aliases: ClC-3; CLC3; CLCN3
UniProt ID: (Human) P51790
Entrez Gene ID: (Human) 1182
Molecular Function: anion channel ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.