Abnova
Manufacturer Code:PAB23850
Catalog # 89-125-558
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human CLC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant CLC.
For Research Use Only
Protein Aliases: Charcot-Leyden crystal protein; CLC; Eosinophil lysophospholipase; Gal-10; galectin 10; Galectin-10; lectin, galactoside-binding, soluble, 10; LGALS10; LPPL_HUMAN; Lysolecithin acylhydrolase; MGC149659
Gene Aliases: CLC; Gal-10; GAL10; LGALS10; LGALS10A; LPPL_HUMAN
UniProt ID: (Human) Q05315
Entrez Gene ID: (Human) 1178
Molecular Function:
cell adhesion molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.