Proteintech
Manufacturer Code:129421AP150UL
Catalog # 50-172-8364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CLASP2 Fusion Protein Ag4012 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLASP2 CLIP associating protein 2 hOrbit2 KIAA0627 Protein Orbit homolog 2; CLASP2alpha; CLASP2gamma; CLIP-associating protein 2; CLIP-associating protein CLASP2; Cytoplasmic linker-associated protein 2; hOrbit2; multiple asters (Mast)-like homolog 2; Protein Orbit homolog 2
Gene Aliases: 1500004F14Rik; 8030404L10Rik; C77448; CLASP2; CLASP2beta; KIAA0627; mKIAA0627
UniProt ID: (Human) O75122, (Mouse) Q8BRT1
Entrez Gene ID: (Human) 23122, (Mouse) 76499
Molecular Function:
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.