Invitrogen
Manufacturer Code:PA530466
Catalog # PIPA530466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 179 of Human CLASP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLIP-associating protein 1; Cytoplasmic linker-associated protein 1; hOrbit1; MAST1 CLIP-associating protein 1 cytoplasmic linker-associated protein 1 multiple asters 1 multiple asters homolog 1 protein Orbit homolog 1; multiple asters 1; Multiple asters homolog 1; Protein Orbit homolog 1
Gene Aliases: 1700030C23Rik; 5730583A19Rik; B130045P17Rik; CLASP1; CLASP1alpha; KIAA0622; MAST1
UniProt ID: (Human) Q7Z460, (Mouse) Q9CVU1
Entrez Gene ID: (Human) 23332, (Mouse) 76707
Molecular Function:
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.