Novus Biologicals
Manufacturer Code:NBP119127
Catalog # NBP119127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1488 and 1538 of human cytoplasmic linker associated protein 1 using the numbering given in entry NP_056097.1 (GeneID 23332). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLIP-associating protein 1; CLIP-associating protein 1 cytoplasmic linker associated protein 1 Cytoplasmic linker-associated protein 1 DKFZp686D1968 DKFZp686H2039 hOrbit1 KIAA0622FLJ33821 MAST1FLJ41222 MGC131895 multiple asters 1 Multiple asters homolog 1 Protein Orbit homolog 1; Cytoplasmic linker-associated protein 1; hOrbit1; multiple asters 1; Multiple asters homolog 1; Protein Orbit homolog 1
Gene Aliases: CLASP1; KIAA0622; MAST1
UniProt ID: (Human) Q7Z460
Entrez Gene ID: (Human) 23332
Molecular Function:
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.