Abnova
Manufacturer Code:H00023332A01
Catalog # 89-106-584
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1133 a.a. to 1226 a.a) partial recombinant protein with GST tag.,CLASP1 (NP_056097 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant CLASP1.
For Research Use Only
Protein Aliases: CLIP-associating protein 1; Cytoplasmic linker-associated protein 1; DKFZp686D1968; DKFZp686H2039; FLJ33821; FLJ41222; hOrbit1; KIAA0622; MAST1; MGC131895; multiple asters 1; Multiple asters homolog 1; Protein Orbit homolog 1
Gene Aliases: CLASP1; KIAA0622; MAST1
UniProt ID: (Human) Q7Z460
Entrez Gene ID: (Human) 23332
Molecular Function:
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.