Invitrogen
Manufacturer Code:PA529341
Catalog # PIPA529341
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 75 and 415 of Human sMtCK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic-type mitochondrial creatine kinase; Creatine kinase S-type, mitochondrial; creatine kinase, mitochondrial 2 (sarcomeric); Mib-CK; S-MtCK; Sarcomeric mitochondrial creatine kinase; SMTCK S-MtCK basic-type mitochondrial creatine kinase creatine kinase S-type mitochondrial mib-CK sarcomeric mitochondrial creatine kinase
Gene Aliases: CKMT2; SMTCK
UniProt ID: (Human) P17540
Entrez Gene ID: (Human) 1160
Molecular Function:
amino acid kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.