Novus Biologicals
Manufacturer Code:NBP292406100UL
Catalog # NB026682
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 40-230 of human CKMT2 (NP_001816.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic-type mitochondrial creatine kinase; Basic-type mitochondrial creatine kinase creatine kinase S-type mitochondrial creatine kinase mitochondrial 2 (sarcomeric) EC 2.7.3 EC 2.7.3.2 mib-CK Sarcomeric mitochondrial creatine kinase S-MtCK SMTCK; Creatine kinase S-type, mitochondrial; creatine kinase, mitochondrial 2 (sarcomeric); Mib-CK; S-MtCK; Sarcomeric mitochondrial creatine kinase
Gene Aliases: CKMT2; SMTCK
UniProt ID: (Human) P17540
Entrez Gene ID: (Human) 1160
Molecular Function:
amino acid kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.