Proteintech
Manufacturer Code:153461AP150UL
Catalog # 50-173-0164
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CKMT1A Fusion Protein Ag7583 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acidic-type mitochondrial creatine kinase; c CKMiMi CKMT CKMT1 CKMT1A Mia CK U MtCK; Creatine kinase U-type, mitochondrial; creatine kinase, mitochondrial 1 (ubiquitous); Mia-CK; U-MtCK; Ubiquitous mitochondrial creatine kinase
Gene Aliases: CKMT; CKMT1; CKMT1A; CKMT1B; mia-CK; U-MtCK
UniProt ID: (Human) P12532
Entrez Gene ID: (Human) 548596
Molecular Function: amino acid kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.