Proteintech
Manufacturer Code:158911AP150UL
Catalog # 50-173-0555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CKM/CKB Fusion Protein Ag8678 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-CK; Brain creatine kinase; CKMM Creatine Kinase Creatine Kinase MM Creatine kinase M chain Creatine kinase M type creatine kinase muscle M CK; CPK-B; CPK-M; Creatine kinase B chain; Creatine kinase B-type; creatine kinase brain; creatine kinase brain-type; Creatine kinase M chain; Creatine kinase M-type; creatine kinase, brain; creatine kinase, muscle; creatine kinase-B; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; M-CK
Gene Aliases: B-CK; BCK; Ck-3; Ck3; CKB; CKBB; Ckbr; CKM; CKMM; HEL-211; HEL-S-29; M-CK; MCK
UniProt ID: (Human) P12277, (Human) P06732, (Rat) P07335, (Mouse) Q04447, (Rat) P00564, (Mouse) P07310
Entrez Gene ID: (Human) 1152, (Human) 1158, (Rat) 24264, (Mouse) 12709, (Rat) 24265, (Mouse) 12715
Molecular Function: amino acid kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.