Abnova
Manufacturer Code:H00001158M06
Catalog # 89-021-094
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2F3-B11 |
Immunogen | CKM (AAH07462 1 a.a. ~ 381 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant CKM.
For Research Use Only
Protein Aliases: CKMM; CPK-M; Creatine kinase M chain; Creatine kinase M-type; creatine kinase, muscle; Creatine phosphokinase M-type; M-CK
Gene Aliases: CKM; CKMM; M-CK
UniProt ID: (Human) P06732
Entrez Gene ID: (Human) 1158
Molecular Function:
amino acid kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.