Novus Biologicals
Manufacturer Code:NBP237457
Catalog # NBP237457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10 |
Immunogen | Purified recombinant fragment of human CSNK2A2 (AA: 194-350) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: casein kinase 2 alpha prime polypeptide casein kinase II subunit alpha' CK II alpha' CK2A2 CSNK2A1 EC 2.7.11 EC 2.7.11.1 FLJ43934; casein kinase 2 alpha'; casein kinase 2, alpha prime polypeptide; Casein kinase II subunit alpha'; CK II alpha'
Gene Aliases: CK2A2; CK2alpha'; CSNK2A1; CSNK2A2
UniProt ID: (Human) P19784
Entrez Gene ID: (Human) 1459
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.