Novus Biologicals
Manufacturer Code:NBP292102100UL
Catalog # NB026096
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 304-423 of human CSNK1G3 (NP_001026982.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: casein kinase 1 gamma 3 casein kinase I isoform gamma-3 CKI-gamma 3 EC 2.7.11 EC 2.7.11.1; casein kinase 1, gamma 3; Casein kinase I isoform gamma-3; CKI-gamma 3
Gene Aliases: CKI-gamma 3; CSNK1G3; CSNK1G3L
UniProt ID: (Human) Q9Y6M4
Entrez Gene ID: (Human) 1456
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.