Invitrogen
Manufacturer Code:MA517062
Catalog # PIMA517062
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E1 |
Immunogen | Purified recombit fragment of human CSNK2A2 (amino acids: 194-350) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: casein kinase 2 alpha'; casein kinase 2, alpha prime polypeptide; Casein kinase II subunit alpha'; casein kinase II, alpha 2, polypeptide; CK II alpha'
Gene Aliases: 1110035J23Rik; C77789; CK2; CK2A2; CK2alpha'; CSNK2A1; CSNK2A2
UniProt ID: (Human) P19784, (Mouse) O54833
Entrez Gene ID: (Human) 1459, (Mouse) 13000, (Rat) 307641
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.