Invitrogen
Manufacturer Code:PA586364
Catalog # PIPA586364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 220-270 of Human Casein Kinase IIalpha. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: casein kinase 2, alpha 1 polypeptide; casein kinase II alpha 1 polypeptide pseudogene; casein kinase II alpha 1 subunit; Casein kinase II subunit alpha; casein kinase II, alpha 1 polypeptide; casein kinase II, alpha 1 related sequence 4; CK II alpha; CK II alpha 3; CK II CKII alpha 1457 CSNK2A1 CK2A1 CKII CKII alpha Casein kinase II subunit alpha Casein Kinase IIalpha CKIIalpha CK-IIalpha CKII-alpha CK IIalpha CKII alpha CKIIalpha; CK2 catalytic subunit alpha; protein kinase CK2
Gene Aliases: CK2A1; CKII; Ckiia; CSNK2A1; Csnk2a1-rs4; CSNK2A3
UniProt ID: (Human) P68400, (Mouse) Q60737, (Rat) P19139
Entrez Gene ID: (Human) 1457, (Mouse) 12995, (Rat) 116549
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.