Invitrogen
Manufacturer Code:PA529185
Catalog # PIPA529185
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 350 of Human CKII alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: casein kinase 2 alpha'; casein kinase 2, alpha 1 polypeptide; casein kinase 2, alpha prime polypeptide; casein kinase II alpha 1 polypeptide pseudogene; casein kinase II alpha 1 subunit; Casein kinase II subunit alpha; Casein kinase II subunit alpha'; CK II alpha; CK II alpha 3; CK II alpha'; CK2 catalytic subunit alpha; CK2A2 CSNK2A1 CK II alpha&prime casein kinase II subunit alpha′ protein kinase CK2
Gene Aliases: CK2A1; CK2A2; CK2alpha'; CKII; CSNK2A1; CSNK2A2; CSNK2A3
UniProt ID: (Human) P68400, (Human) P19784
Entrez Gene ID: (Human) 1457, (Human) 1459
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.