Invitrogen
Manufacturer Code:PA532129
Catalog # PIPA532129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 144 and 361 of Human Casein Kinase 1 delta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: casein kinase 1, delta; Casein kinase I isoform delta; CKI-delta; CKId; CKIdelta HCKID CKI-delta CKId casein kinase I isoform delta tau-protein kinase CSNK1D; Tau-protein kinase CSNK1D
Gene Aliases: 1200006A05Rik; AA409348; ASPS; CKIdelta; CSNK1D; D930010H05Rik; FASPS2; HCKID
UniProt ID: (Human) P48730, (Mouse) Q9DC28
Entrez Gene ID: (Human) 1453, (Mouse) 104318
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.