Invitrogen
Manufacturer Code:PA534462
Catalog # PIPA534462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid synthetic peptide near the amino terminus of human CITED2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cbp/p300-interacting transactivator 2; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 2; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2; CITED2 Cbp/p300-interacting transactivator 2 melanocyte-specific gene 1-related gene 1 MSG-related protein 1 MRG1 melanocyte specific gene 2 MSG2 ER154-like ASD8 VSD2; melanocyte specific gene 2; melanocyte-specific gene 1-related gene 1; MRG-1; MSG-related protein 1; MSG1-related gene 1; P35srj
Gene Aliases: AI835299; ASD8; CITED2; ER154-like; MRG-1; MRG1; Msg2; P35SRJ; VSD2
UniProt ID: (Human) Q99967, (Mouse) O35740
Entrez Gene ID: (Human) 10370, (Rat) 114490, (Mouse) 17684
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.