Novus Biologicals
Manufacturer Code:NBP271199R
Catalog # NBP271199R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3C12 |
Immunogen | Full length recombinant protein of human CITED1 (NP_004134) produced in E.coli. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Cbp/p300-interacting transactivator 1; cbp/p300-interacting transactivator 1 Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain1MSG1Melanocyte-specific protein 1 melanocyte-specific gene 1; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 1; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 1; melanocyte-specific gene 1; Melanocyte-specific protein 1
Gene Aliases: CITED1; MSG1
UniProt ID: (Human) Q99966
Entrez Gene ID: (Human) 4435
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.