Invitrogen
Manufacturer Code:MA527296
Catalog # PIMA527296
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4D12 |
Immunogen | Full length human recombit protein of CISD2 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CDGSH iron-sulfur domain-containing protein 2; CISD2 CDGSH iron-sulfur domain-containing protein 2 endoplasmic reticulum intermembrane small protein ERIS mitoNEET related 1 miner1 nutrient-deprivation autophagy factor-1 NAF-1 zinc finger CDGSH-type domain 2 ZCD2 WFS2; Endoplasmic reticulum intermembrane small protein; Miner1; MitoNEET-related 1 protein; NAF-1; Nutrient-deprivation autophagy factor-1; zinc finger, CDGSH-type domain 2
Gene Aliases: CDGSH2; CISD2; ERIS; Miner1; NAF-1; WFS2; ZCD2
UniProt ID: (Human) Q8N5K1
Entrez Gene ID: (Human) 493856
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.