Novus Biologicals
Manufacturer Code:NBP159676
Catalog # NBP159676
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to CISD2(CDGSH iron sulfur domain 2) The peptide sequence was selected from the N terminal of CISD2. Peptide sequence VLESVARIVKVQLPAYLKRLPVPESITGFARLTVSEWLRLLPFLGVLALL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDGSH iron sulfur domain 2 CDGSH iron-sulfur domain-containing protein 2 CDGSH2 Endoplasmic reticulum intermembrane small protein ERISWFS2 Miner1Wolfram syndrome 2 mitoNEET related 1 MitoNEET-related 1 protein NAF-1 Nutrient-deprivation autophagy factor-1 ZCD2CDGSH iron sulfur domain-containing protein 2 zinc finger CDGSH-type domain 2; CDGSH iron-sulfur domain-containing protein 2; Endoplasmic reticulum intermembrane small protein; Miner1; MitoNEET-related 1 protein; NAF-1; Nutrient-deprivation autophagy factor-1; zinc finger, CDGSH-type domain 2
Gene Aliases: CDGSH2; CISD2; ERIS; Miner1; NAF-1; WFS2; ZCD2
UniProt ID: (Human) Q8N5K1
Entrez Gene ID: (Human) 493856
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.