Proteintech
Manufacturer Code:160061AP150UL
Catalog # 50-173-0637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CISD1 Fusion Protein Ag8680 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C10orf70 CDGSH iron sulfur domain 1 CISD1 MDS029 mitoNEET Mnt ZCD1; CDGSH iron sulfur domain-containing protein 1; CDGSH iron-sulfur domain-containing protein 1; MitoNEET; zinc finger CDGSH-type domain 1; zinc finger, CDGSH-type domain 1
Gene Aliases: AU043990; AW743335; C10orf70; CISD1; D10Ertd214e; MDS029; MitoNEET; RGD1309529; si:dkey-18n13.4; ZCD1; zgc:63561
UniProt ID: (Human) Q9NZ45, (Mouse) Q91WS0, (Rat) B0K020
Entrez Gene ID: (Human) 55847, (Mouse) 52637, (Rat) 294362, (Zebrafish) 393577
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.