Invitrogen
Manufacturer Code:MA516081
Catalog # PIMA516081
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G10 |
Immunogen | A C-terminal portion of the human p90 protein. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cancerous inhibitor of PP2A; cancerous inhibitor of protein phosphatase 2A; CIP2A p90 cancerous inhibitor of PP2A p90 autoantigen protein CIP2A; p90 autoantigen; p90 autoantigen homolog; Protein CIP2A
Gene Aliases: AA408511; AU018569; CIP2A; KIAA1524; p90
UniProt ID: (Mouse) Q8BWY9, (Human) Q8TCG1
Entrez Gene ID: (Mouse) 224171, (Human) 57650
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.