Invitrogen
Manufacturer Code:PA572334
Catalog # PIPA572334
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 269-301 amino acids from human KIAA1524 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancerous inhibitor of PP2A; p90 autoantigen homolog; Protein CIP2A; Protein CIP2A Cancerous inhibitor of PP2A p90 autoantigen KIAA1524 CIP2A
Gene Aliases: AA408511; AU018569; Cip2a; Kiaa1524
UniProt ID: (Mouse) Q8BWY9
Entrez Gene ID: (Mouse) 224171
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.