Novus Biologicals
Manufacturer Code:NBP29297520UL
Catalog # NB027774
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 681-905 of human KIAA1524 (NP_065941.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancerous inhibitor of PP2A; Cancerous inhibitor of PP2A CIP2A FLJ12850 KIAA1524 MGC163436 p90 p90 autoantigen protein CIP2A; cancerous inhibitor of protein phosphatase 2A; p90 autoantigen; Protein CIP2A
Gene Aliases: CIP2A; KIAA1524; p90
UniProt ID: (Human) Q8TCG1
Entrez Gene ID: (Human) 57650
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.