Invitrogen
Manufacturer Code:PA568760
Catalog # PIPA568760
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of mouse CIITA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C2TA CIITAIV MHC2TA NLRA MHC class II transactivator MHC class II transactivator type III NLR family acid domain containing nucleotide-binding oligomerization domain leucine rich repeat and acid domain containing; CIITA; class II transactivator; MHC class II transactivator; MHC class II transactivator type I; MHC class II transactivator type III; MHC class II transactivator type IV; NLR family, acid domain containing; nucleotide-binding oligomerization domain, leucine rich repeat and acid domain containing
Gene Aliases: C2TA; CIITA; CIITAIV; EG669998; Gm9475; MHC2TA; NLRA
UniProt ID: (Human) P33076, (Mouse) P79621
Entrez Gene ID: (Human) 4261, (Dog) 490008, (Mouse) 12265, (Rat) 85483
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.