Invitrogen
Manufacturer Code:PA5101105
Catalog # PIPA5101105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CIITA corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C2ta CIITA CIITAIV class II transactivator class II major histocompatibility complex transactivator EG669998 Gm9475 MH NLRA MHC class II transactivator MHC class II transactivator type I MHC class II transactivator type III MHC class II transactivator type IV MHC2TA NLR family acid domain containing NLRA nucleotide-binding oligomerization domain leucine rich repeat and acid domain containing; CIITA; class II transactivator; MHC class II transactivator; MHC class II transactivator type I; MHC class II transactivator type III; MHC class II transactivator type IV; NLR family, acid domain containing; nucleotide-binding oligomerization domain, leucine rich repeat and acid domain containing
Gene Aliases: C2TA; CIITA; CIITAIV; EG669998; Gm9475; MHC2TA; NLRA
UniProt ID: (Human) P33076, (Mouse) P79621
Entrez Gene ID: (Human) 4261, (Mouse) 12265, (Rat) 85483
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.