Novus Biologicals
Manufacturer Code:NBP259074
Catalog # NBP259074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Anti-CIITA was produced by repeated immunizations with CIITA peptide corresponding to a region near the N-terminus of the human protein conjugated to Keyhole Limpet Hemocyanin (KLH). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C2TA C2TAMHC class II transactivator type III CIITAIV class II major histocompatibility complex transactivator MHC class II transactivator MHC2TANLR family acid domain containing NLRA nucleotide-binding oligomerization domain leucine rich repeat and acid domaincontaining; CIITA; MHC class II transactivator; MHC class II transactivator type I; MHC class II transactivator type III; NLR family, acid domain containing; nucleotide-binding oligomerization domain, leucine rich repeat and acid domain containing
Gene Aliases: C2TA; CIITA; CIITAIV; MHC2TA; NLRA
UniProt ID: (Human) P33076
Entrez Gene ID: (Human) 4261
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.