Abnova
Manufacturer Code:PAB19313
Catalog # 89-120-846
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 16 amino acids near N-terminus of human CIITA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody rasied against synthetic peptide of CIITA.
For Research Use Only
Protein Aliases: C2TA; CIITA; CIITAIV; MHC class II transactivator; MHC class II transactivator type I; MHC class II transactivator type III; MHC2TA; NLR family, acid domain containing; NLRA; nucleotide-binding oligomerization domain, leucine rich repeat and acid domain containing
Gene Aliases: C2TA; CIITA; CIITAIV; MHC2TA; NLRA
UniProt ID: (Human) P33076
Entrez Gene ID: (Human) 4261
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.