Invitrogen
Manufacturer Code:PA5103387
Catalog # PIPA5103387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CIDEC corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell death activator CIDE-3; Cell death activator CIDE-3 cell death inducing DFFA like effector c cell death-inducing DFFA-like effector c cell death-inducing DFFA-like effector protein C CID cide 3 CIDE3 CIDE-3 CIDE-3alpha Cidec fat specific gene 27 fat specific protein 27 fat-specific protein FSP27 Fat-specific protein FSP27 homolog FPLD5 Fsp27; cell death-inducing DFFA-like effector c; Cell death-inducing DFFA-like effector protein C; fat specific protein 27; Fat-specific protein FSP27 homolog
Gene Aliases: CIDE-3; CIDE3; CIDEC; FPLD5; FSP27
UniProt ID: (Human) Q96AQ7
Entrez Gene ID: (Human) 63924
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.