Novus Biologicals
Manufacturer Code:NB100430
Catalog # NB100430
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Residues 10-25 LLYPKSLSRHVSVRTS] of the 27 kDa human FSP27 (CIDE-3) protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell death activator CIDE-3; cell death activator CIDE-3 cell death-inducing DFFA-like effector c Cell death-inducing DFFA-like effector protein C CIDE-3 CIDEX Fat-specific protein FSP27 homolog FLJ20871 Fsp27 FSP27fat specific protein 27; cell death-inducing DFFA-like effector c; Cell death-inducing DFFA-like effector protein C; fat specific protein 27; Fat-specific protein FSP27 homolog
Gene Aliases: CIDE-3; CIDE3; CIDEC; FPLD5; FSP27
UniProt ID: (Human) Q96AQ7
Entrez Gene ID: (Human) 63924
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.