Abnova
Manufacturer Code:H00063924B01P
Catalog # 89-017-145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | CIDEC (AAH16851 1 a.a. ~ 238 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human CIDEC protein.
For Research Use Only
Protein Aliases: Cell death activator CIDE-3; cell death-inducing DFFA-like effector c; Cell death-inducing DFFA-like effector protein C; CIDE-3; fat specific protein 27; Fat-specific protein FSP27 homolog; FLJ20871; Fsp27
Gene Aliases: CIDE-3; CIDE3; CIDEC; FPLD5; FSP27
UniProt ID: (Human) Q96AQ7
Entrez Gene ID: (Human) 63924
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.