Novus Biologicals
Manufacturer Code:NBP176951
Catalog # NBP176951
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 18 amino acid peptide near the carboxy terminus of human CIDE-A. The immunogen is located within the last 50 amino acids of CIDE-A. Amino Acid Squence: DDKEERPSLRSQAKGRFT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell death activator CIDE-A; cell death activator CIDE-A cell death-inducing DFFA-like effector aCIDE-A; Cell death-inducing DFFA-like effector A
Gene Aliases: CIDE-A; CIDEA
UniProt ID: (Human) O60543
Entrez Gene ID: (Human) 1149
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.