Invitrogen
Manufacturer Code:MA538025
Catalog # PIMA538025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2354 |
Immunogen | A synthesized peptide derived from human CHX10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: C. elegans ceh-10 homeo domain containing homolog; C. elegans ceh-10 homeo domain containing homolog ceh-10 homeo domain containing homolog ceh-10 homeodomain containing homolog ceh-10 homeodomain-containing homolog CHX10 Homeobox protein CHX10 HOX10 Hox-10 MCOP2 MCOPCB3 ocular retardation or RET1 Visual system homeobox 2 Vsx2; ceh-10 homeo domain containing homolog; Ceh-10 homeodomain-containing homolog; Homeobox protein CHX10; ocular retardation; Visual system homeobox 2
Gene Aliases: CHX10; Hox-10; HOX10; MCOP2; MCOPCB3; or; RET1; VSX2
UniProt ID: (Human) P58304, (Mouse) Q61412
Entrez Gene ID: (Human) 338917, (Mouse) 12677, (Rat) 171360
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.