Invitrogen
Manufacturer Code:PA585404
Catalog # PIPA585404
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human CHX10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C. elegans ceh-10 homeo domain containing homolog; Ceh-10 homeodomain-containing homolog; CHX10 antibody HOX10 antibody MCOP2 antibody MCOPCB3 antibody RET1 antibody VSX2 antibody ceh-10 homeo domain containing homolog antibody homeobox protein CHX10 antibody visual system homeobox 2 antibody ceh-10 homeodomain-containing homolog antibody; Homeobox protein CHX10; ocular retardation; Visual system homeobox 2
Gene Aliases: Chx10; Hox-10; Hox10; or; Vsx2
UniProt ID: (Mouse) Q61412
Entrez Gene ID: (Mouse) 12677
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.