Invitrogen
Manufacturer Code:PA131287
Catalog # PIPA131287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fragment corresponding to N terminal amino acids 1-131 of Human CHX10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: ceh-10 homeo domain containing homolog; Ceh-10 homeodomain-containing homolog; CHX10 HOX10 MCOP2 MCOPCB3 RET1; Homeobox protein CHX10; Visual system homeobox 2
Gene Aliases: CHX10; HOX10; MCOP2; MCOPCB3; RET1; VSX2
UniProt ID: (Human) P58304
Entrez Gene ID: (Human) 338917
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.