Novus Biologicals
Manufacturer Code:NBP2846800.1ML
Catalog # NB032608
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of human CHX10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ceh-10 homeo domain containing homolog; ceh-10 homeo domain containing homolog ceh-10 homeo domain containing homolog (C. elegans) ceh-10 homeodomain containing homolog (C. elegans) Ceh-10 homeodomain-containing homolog CHX10MCOPCB3 Homeobox protein CHX10 HOX10C elegans ceh-10 homeo domain-containing homolog MCOP2 RET1 visual system homeobox 2; Ceh-10 homeodomain-containing homolog; Homeobox protein CHX10; Visual system homeobox 2
Gene Aliases: CHX10; HOX10; MCOP2; MCOPCB3; RET1; VSX2
UniProt ID: (Human) P58304
Entrez Gene ID: (Human) 338917
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.