Invitrogen
Manufacturer Code:PA544803
Catalog # PIPA544803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CHTF18 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C321D2.3 (novel protein); C321D2.4 (novel protein); CHL12; Chl12 homolog; Chromosome transmission fidelity protein 18 homolog; CTF18 Chromosome transmission fidelity factor 18 homolog (S. cerevisiae) C16orf41 C321D2.2 C321D2.3 C321D2.4 CHL12 Ctf18 RUVBL; CTF18, chromosome transmission fidelity factor 18 homolog; hCTF18; homolog of yeast CHL12; some homology with holliday junction DNA helicase RUVB like
Gene Aliases: 6030457M03Rik; C16orf41; C321D2.2; C321D2.3; C321D2.4; CHL12; CHTF18; CTF18; RUVBL; zgc:113153
UniProt ID: (Human) Q8WVB6, (Mouse) Q8BIW9
Entrez Gene ID: (Human) 63922, (Horse) 100067225, (Pig) 100520370, (Bovine) 517007, (Dog) 490084, (Mouse) 214901, (Rat) 287146, (Guinea pig) 100722938, (Zebrafish) 554366
Molecular Function: DNA binding protein hydrolase nucleic acid binding nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.