Novus Biologicals
Manufacturer Code:NBP157605
Catalog # NBP157605
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to CHTF18 (CTF18 chromosome transmission fidelity factor 18 homolog (S. cerevisiae)) The peptide sequence was selected from the middle region of CHTF18)(50ug). Peptide sequence QALLLDALCLLLDILAPKLRPVSTQLYST |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C16orf41 C321D2.2 C321D2.3 C321D2.3 (novel protein) C321D2.4 (novel protein) CHL12C321D2.4 chromosome 16 open reading frame 41 chromosome transmission fidelity protein 18 homolog CTF18 CTF18 chromosome transmission fidelity factor 18 homolog (S. cerevisiae) EC 3.6.1 hCTF18 homolog of yeast CHL12 RUVBL some homology with holliday junction DNA helicase RUVB like; C321D2.3 (novel protein); C321D2.4 (novel protein); CHL12; Chromosome transmission fidelity protein 18 homolog; CTF18, chromosome transmission fidelity factor 18 homolog; hCTF18; homolog of yeast CHL12; some homology with holliday junction DNA helicase RUVB like
Gene Aliases: C16orf41; C321D2.2; C321D2.3; C321D2.4; CHL12; CHTF18; CTF18; RUVBL
UniProt ID: (Human) Q8WVB6
Entrez Gene ID: (Human) 63922
Molecular Function: DNA binding protein hydrolase nucleic acid binding nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.