Novus Biologicals
Manufacturer Code:NB11074570SS
Catalog # NB11074570T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 62-2E8 |
Immunogen | Recombinant fusion protein from the C-terminal portion of the human CHT1 protein. [UniProt# Q9GZV3] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CHT; CHT CHT1MGC126299 hCHT Hemicholinium-3-sensitive choline transporter high affinity choline transporter 1 high affinity choline transporter hemicholinium-3-sensitive cholinetransporter MGC126300 solute carrier family 5 (choline transporter) member 7 Solute carrier family 5 member 7; Hemicholinium-3-sensitive choline transporter; High affinity choline transporter 1; high affinity choline transporter; hemicholinium-3-sensitive choline transporter; solute carrier family 5 (sodium/choline cotransporter), member 7; Solute carrier family 5 member 7
Gene Aliases: CHT; CHT1; HMN7A; SLC5A7
UniProt ID: (Human) Q9GZV3
Entrez Gene ID: (Human) 60482
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.