Novus Biologicals
Manufacturer Code:NBP2866040.1ML
Catalog # NB034228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human CHT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHT; CHT CHT1MGC126299 hCHT Hemicholinium-3-sensitive choline transporter high affinity choline transporter 1 high affinity choline transporter hemicholinium-3-sensitive cholinetransporter MGC126300 solute carrier family 5 (choline transporter) member 7 Solute carrier family 5 member 7; Hemicholinium-3-sensitive choline transporter; High affinity choline transporter 1; high affinity choline transporter; hemicholinium-3-sensitive choline transporter; solute carrier family 5 (sodium/choline cotransporter), member 7; Solute carrier family 5 member 7
Gene Aliases: CHT; CHT1; HMN7A; SLC5A7
UniProt ID: (Human) Q9GZV3
Entrez Gene ID: (Human) 60482
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.