Invitrogen
Manufacturer Code:PA572204
Catalog # PIPA572204
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 295-329 amino acids from the Central region of human CHSY3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carbohydrate synthase 2; Chondroitin glucuronyltransferase 3; Chondroitin sulfate synthase 3; Chondroitin sulfate synthase 3 Carbohydrate synthase 2 Chondroitin glucuronyltransferase 3 Chondroitin synthase 2 ChSy-2 Glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase II N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase 3 N-acetylgalactosaminyltransferase 3 CHSY3 CHSY2 CSS3; Chondroitin synthase 2; chondroitin synthase-2; ChSy-2; Glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase II; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase 3; N-acetylgalactosaminyltransferase 3
Gene Aliases: 4833446K15Rik; AI662215; CHSY2; CHSY3; CSS3; Kiaa4168
UniProt ID: (Human) Q70JA7, (Mouse) Q5DTK1
Entrez Gene ID: (Human) 337876, (Mouse) 78923
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.