Novus Biologicals
Manufacturer Code:NBP2846780.1ML
Catalog # NB032606
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human CHSS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carbohydrate synthase 1; Chondroitin glucuronyltransferase 1; Chondroitin glucuronyltransferase 1 chondroitin glucuronyltransferase II chondroitin sulfate synthase 1 Chondroitin synthase 1 CHSY ChSy-1 CSS1DKFZp434M032 EC 2.4.1.175 EC 2.4.1.226 FLJ58581 Glucuronosyl-N-acetylgalactosaminyl-proteoglycan4-beta-N-acetylgalactosaminyltransferase 1 KIAA0990carbohydrate (chondroitin) synthase 1 N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase 1 N-acetylgalactosaminyltransferase 1 N-acetylgalactosaminyltransferase II; chondroitin glucuronyltransferase II; Chondroitin sulfate synthase 1; Chondroitin synthase 1; ChSy-1; Glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase 1; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase 1; N-acetylgalactosaminyltransferase 1; N-acetylgalactosaminyltransferase II
Gene Aliases: CHSY; ChSy-1; CHSY1; CSS1; KIAA0990; TPBS; UNQ756/PRO1487
UniProt ID: (Human) Q86X52
Entrez Gene ID: (Human) 22856
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.