Proteintech
Manufacturer Code:144201AP150UL
Catalog # 50-172-9438
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHSY1 Fusion Protein Ag5766 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carbohydrate (chondroitin) synthase 1; carbohydrate synthase 1; Chondroitin glucuronyltransferase 1; chondroitin glucuronyltransferase II; Chondroitin sulfate synthase 1; chondroitin sulfate synthase 1 Chondroitin synthase 1 CHSY ChSy 1 CHSY1 CSS1 KIAA0990; Chondroitin synthase 1; ChSy-1; Glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase 1; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase 1; N-acetylgalactosaminyltransferase 1; N-acetylgalactosaminyltransferase II
Gene Aliases: CHSY; ChSy-1; CHSY1; chys1; CSS1; KIAA0990; mKIAA0990; TPBS; UNQ756/PRO1487; wu:fc27h05; wu:fc27h06; zgc:64191
UniProt ID: (Human) Q86X52, (Mouse) Q6ZQ11
Entrez Gene ID: (Human) 22856, (Mouse) 269941, (Rat) 292999, (Zebrafish) 324407
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.