Invitrogen
Manufacturer Code:PA530842
Catalog # PIPA530842
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 200 and 435 of Human CHST9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9; Carbohydrate sulfotransferase 9; GalNAc-4-O-sulfotransferase 2; GalNAc-4-ST2; GalNAc-4-sulfotransferase 2; GalNAc4ST-2; GALNAC4ST-2 GalNAc-4-sulfotransferase 2 N-acetylgalactosamine 4-O-sulfotransferase 2 carbohydrate sulfotransferase 9 galNAc-4-O-sulfotransferase 2; N-acetylgalactosamine 4-O-sulfotransferase 2; N-acetylgalactosamine-4-O-sulfotransferase 2
Gene Aliases: 5430438D01Rik; CHST9; GALNAC4ST-2; GalNAc4ST2; UNQ2549/PRO6175
UniProt ID: (Human) Q7L1S5, (Mouse) Q76EC5
Entrez Gene ID: (Human) 83539, (Mouse) 71367
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.