Invitrogen
Manufacturer Code:PA5113021
Catalog # PIPA5113021
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Carbohydrate sulfotransferase 9 protein (33-129AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5430438D01Rik carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9 Carbohydrate sulfotransferase 9 CHST9 GalNAc-4-O-sulfotransferase 2 GalNAc4ST2 GalNAc-4-ST2 GALNAC4ST-2 GalNAc-4-sulfotransferase 2 N-acetylgalactosamine 4-O-sulfotransferase 2 N-acetylgalactosamine-4-O-sulfotransferase 2 UNQ2549/PRO6175; carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9; Carbohydrate sulfotransferase 9; GalNAc-4-O-sulfotransferase 2; GalNAc-4-ST2; GalNAc-4-sulfotransferase 2; GalNAc4ST-2; N-acetylgalactosamine 4-O-sulfotransferase 2; N-acetylgalactosamine-4-O-sulfotransferase 2
Gene Aliases: 5430438D01Rik; CHST9; GALNAC4ST-2; GalNAc4ST2; UNQ2549/PRO6175
UniProt ID: (Human) Q7L1S5, (Mouse) Q76EC5
Entrez Gene ID: (Human) 83539, (Mouse) 71367
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.