Novus Biologicals
Manufacturer Code:NBP215898
Catalog # NBP215898
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human CHST9. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9; carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9 carbohydrate sulfotransferase 9 EC 2.8.2.- FLJ42328 GalNAc-4-O-sulfotransferase 2 GalNAc-4-ST2 GALNAC4ST-2 GalNAc-4-sulfotransferase 2 N-acetylgalactosamine 4-O-sulfotransferase 2 N-acetylgalactosamine-4-O-sulfotransferase 2; Carbohydrate sulfotransferase 9; GalNAc-4-O-sulfotransferase 2; GalNAc-4-ST2; GalNAc-4-sulfotransferase 2; N-acetylgalactosamine 4-O-sulfotransferase 2; N-acetylgalactosamine-4-O-sulfotransferase 2
Gene Aliases: CHST9; GALNAC4ST-2; GalNAc4ST2; UNQ2549/PRO6175
UniProt ID: (Human) Q7L1S5
Entrez Gene ID: (Human) 83539
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.