Invitrogen
Manufacturer Code:PA543412
Catalog # PIPA543412
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CHST8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 8; Carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 8 GalNAc4ST GALNAC4ST1; carbohydrate N-acetylgalactosamine 4-0 sulfotransferase 8; Carbohydrate sulfotransferase 8; GalNAc-4-O-sulfotransferase 1; GALNAC-4-ST1; GalNAc4ST-1; N-acetylgalactosamine-4-O-sulfotransferase 1
Gene Aliases: 1500011J21Rik; AI426009; CHST8; GalNAc4ST; GALNAC4ST1; PSS3
UniProt ID: (Human) Q9H2A9, (Mouse) Q8BQ86
Entrez Gene ID: (Dog) 484596, (Human) 64377, (Horse) 100050006, (Bovine) 100271926, (Rabbit) 100355847, (Rat) 308511, (Guinea pig) 100721873, (Mouse) 68947
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.