Abnova
Manufacturer Code:H00064377K
Catalog # 89-208-804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human CHST8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human CHST8 peptide using ARM Technology.
For Research Use Only
Protein Aliases: carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 8; Carbohydrate sulfotransferase 8; GalNAc-4-O-sulfotransferase 1; GALNAC-4-ST1; galNAc4ST-1; GALNAC4ST1 GalNAc4ST; N-acetylgalactosamine-4-O-sulfotransferase 1
Gene Aliases: CHST8; GalNAc4ST; GALNAC4ST1; PSS3
UniProt ID: (Human) Q9H2A9
Entrez Gene ID: (Human) 64377
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.